6-Brom-[1,2,4]triazolo[4,3-a]pyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-[1,2,4]triazolo[4,3-a]pyridine
OR3548-250MG
OR3548-500MG
APOSOR3548-250MGEA
21.9
EUR
APOSOR3548-250MG
APOSOR3548-500MG
6-Brom-[1,2,4]triazolo[4,3-a]pyridin 98%
6-Brom-[1,2,4]triazolo[4,3-a]pyridin
Formel:
C₆H₄BrN₃ Molecular Weight: 198.0200 g/mol Schmelzpunkt: 151…153 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08689506 CAS-Nummer: 108281-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...