Methyl-5-methyl-4-oxazolcarboxylat 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-methyloxazole-4-carboxylate
OR3554-1G
OR3554-5G
APOSOR3554-1GEA
81.2
EUR
APOSOR3554-1G
APOSOR3554-5G
Methyl-5-methyl-4-oxazolcarboxylat 97%
Methyl-5-methyl-4-oxazolcarboxylat
Formel:
C₆H₇NO₃ Molecular Weight: 141.1247 g/mol Siedepunkt: 51…57 °C/0.1mmHg Schmelzpunkt: 45…48 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08689502 CAS-Nummer: 41172-57-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...