5-Brom-2-chlorpyrimidin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR3560-25G
OR3560-100G
OR3560-500G
APOSOR3560-25GEA
21.9
EUR
APOSOR3560-25G
APOSOR3560-100G
APOSOR3560-500G
5-Brom-2-chlorpyrimidin 98%
5-Bromo-2-chloropyrimidine
Formel:
C₄H₂BrClN₂ Molecular Weight: 193,43 g/mol Siedepunkt: 95 °C/15mmHg Schmelzpunkt: 73…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00483232 CAS-Nummer: 32779-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...