Methyl-2-methoxy-5-sulfamoylbenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Aminosulphonyl)-2-methoxybenzoic acid Methyl Ester, 2-Methoxy-5-sulphamoylbenzoic acid Methyl Ester, Methyl 2-Methoxy-5-sulphamoylbenzoate, Methyl 2-methoxy-5-sulphamoylbenzoate, Sulpiride impurity B

OR3568-100G OR3568-25G
APOSOR3568-100GEA 49.3 EUR
APOSOR3568-100G APOSOR3568-25G
Methyl-2-methoxy-5-sulfamoylbenzoat
Methyl-2-methoxy-5-sulfamoylbenzoat
Formel: C₉H₁₁NO₅S
Molecular Weight: 245,25 g/mol
Schmelzpunkt: 175…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01317542
CAS-Nummer: 33045-52-2
EINECS: 251-358-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR