2-Chlor-6-methyl-5-nitropyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chlor-2-methyl-3-nitropyridin, 2-Chlor-5-nitro-6-picolin, 6-Chlor-3-nitro-2-picolin
OR3578-25G
OR3578-5G
APOSOR3578-25GEA
29
EUR
APOSOR3578-25G
APOSOR3578-5G
2-Chlor-6-methyl-5-nitropyridin 98%
2-Chloro-6-methyl-5-nitropyridine
Formel:
C₆H₅ClN₂O₂ Molecular Weight: 172,57 g/mol Schmelzpunkt: 54…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234160 CAS-Nummer: 22280-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...