4-Benzyl-3-thiosemicarbazid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Benzylhydrazinecarbothioamide
OR3584-1G
OR3584-5G
APOSOR3584-1GEA
59.6
EUR
APOSOR3584-1G
APOSOR3584-5G
4-Benzyl-3-thiosemicarbazid 95%
4-Benzyl-3-thiosemicarbazid
Formel:
C₈H₁₁N₃S Molecular Weight: 181.26 g/mol Schmelzpunkt: 128…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025145 CAS-Nummer: 13431-41-9 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...