2,5-Dichlorbenzophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2,5-Dichlorophenyl)(phenyl)methanone
OR3586-100G
OR3586-25G
APOSOR3586-100GEA
58
EUR
APOSOR3586-100G
APOSOR3586-25G
2,5-Dichlorbenzophenon 98%
2,5-Dichlorbenzophenon
Formel:
C₁₃H₈Cl₂O Molecular Weight: 251,11 g/mol Schmelzpunkt: 88…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079746 CAS-Nummer: 16611-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...