5-(Methylsulphonyl)pyridine-3-boronic acid 98%

Lieferant: Apollo Scientific
Achtung

OR3598-1G OR3598-250MG
APOSOR3598-1GEA 0 EUR
APOSOR3598-1G APOSOR3598-250MG
5-(Methylsulphonyl)pyridine-3-boronic acid 98%
5-(Methylsulphonyl)pyridine-3-boronic acid
Formel: C₆H₈BNO₄S
Molecular Weight: 201,00 g/mol
Schmelzpunkt: 123…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689536
CAS-Nummer: 913836-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR