(4-(Methoxymethoxy)phenyl)boronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Methoxymethoxy)phenylboronic acid, 4-(Methoxymethoxy)benzeneboronic acid

OR360049-25G OR360049-5G
APOSOR360049-25GEA 857 EUR
APOSOR360049-25G APOSOR360049-5G
(4-(Methoxymethoxy)phenyl)boronic acid 98%
(4-(Methoxymethoxy)phenyl)boronic acid
Formel: C₈H₁₁BO₄
Molecular Weight: 181,98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03412067
CAS-Nummer: 162662-27-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR