(5-Ethylpyridin-3-yl)boronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR360079-1G
OR360079-250MG
APOSOR360079-1GEA
406
EUR
APOSOR360079-1G
APOSOR360079-250MG
(5-Ethylpyridin-3-yl)boronic acid 98%
(5-Ethylpyridin-3-yl)boronic acid
Formel:
C₇H₁₀BNO₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
1001907-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...