Deoxyfuconojirimycin hydrochloride
Lieferant: Apollo Scientific
Achtung
Synonyme: (2S,3R,4S,5R)-2-Methyl-3,4,5-trihydroxypiperidine hydrochloride
OR3600T-5MG OR3600T-100MG OR3600T-10MG
APOSOR3600T-5MGEA 219 EUR
APOSOR3600T-5MG APOSOR3600T-100MG APOSOR3600T-10MG
Deoxyfuconojirimycin hydrochloride
Deoxyfuconojirimycin hydrochloride
A powerful & specific inhibitor of several a-L-fucosidases.
Formel: C₆H₁₃NO₃·HCl
Molecular Weight: 183,6 g/mol
Schmelzpunkt: 155…157 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 210174-73-5