2,6-Dimethyl-4-pyridinboronsäurepinakolester

Lieferant: Apollo Scientific

Synonyme: 2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

OR360125-1G
APOSOR360125-1GEA 65.4 EUR
APOSOR360125-1G
2,6-Dimethyl-4-pyridinboronsäurepinakolester
2,6-Dimethyl-4-pyridinboronsäurepinakolester
Formel: C₁₃H₂₀BNO₂
Molecular Weight: 233,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11617853
CAS-Nummer: 325142-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR