4-(4-Methylpiperazin-1-yl)phenylboronic acid 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Methyl-1-piperazinyl)phenylboronic Acid

OR360145-1G OR360145-10G OR360145-5G
APOSOR360145-1GEA 43.5 EUR
APOSOR360145-1G APOSOR360145-10G APOSOR360145-5G
4-(4-Methylpiperazin-1-yl)phenylboronic acid 96%
4-(4-Methylpiperazin-1-yl)phenylboronic acid
Formel: C₁₁H₁₇BN₂O₂
Molecular Weight: 220,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11223486
CAS-Nummer: 229009-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR