4-(2-Naphthyl)phenylboronsäure 95+%

Lieferant: Apollo Scientific

Synonyme: (4-(Naphthalen-2-yl)phenyl)boronic acid

OR360155-25G
APOSOR360155-25GEA 194 EUR
APOSOR360155-25G
4-(2-Naphthyl)phenylboronsäure 95+%
4-(2-Naphthyl)phenylboronsäure
Formel: C₁₆H₁₃BO₂
Molecular Weight: 248.0800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260454
CAS-Nummer: 918655-03-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR