(4-Chlorocarbonylphenyl)boronic acid

Lieferant: Apollo Scientific

Synonyme: Benzoylchloride-4-boronic acid, (4-(Chlorocarbonyl)phenyl)boronic acid, 4-Chlorocarbonylphenylboronic boronic acid

OR360238-1G OR360238-25G OR360238-5G
APOSOR360238-1GEA 87 EUR
APOSOR360238-1G APOSOR360238-25G APOSOR360238-5G
(4-Chlorocarbonylphenyl)boronic acid
(4-Chlorocarbonylphenyl)boronic acid
Formel: C₇H₆BClO₃
Molecular Weight: 184.38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08062416
CAS-Nummer: 332154-57-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR