2-Brom-4-pyridinboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Brompyridin-4-boronsäure, 2-Brom-4-pyridinylboronsäure
OR360326-5G
APOSOR360326-5GEA
252
EUR
APOSOR360326-5G
2-Brom-4-pyridinboronsäure 98%
2-Bromo-4-pyridineboronic acid
Formel:
C₅H₅BBrNO₂ Molecular Weight: 201,81 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06798244 CAS-Nummer: 458532-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...