4-Brom-2-methoxyphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Bromo-2-methoxyphenyl)boronic acid
OR360478-1G
OR360478-5G
OR360478-10G
APOSOR360478-1GEA
60.9
EUR
APOSOR360478-1G
APOSOR360478-5G
APOSOR360478-10G
4-Brom-2-methoxyphenylboronsäure 98%
4-Brom-2-methoxyphenylboronsäure
Formel:
C₇H₈BBrO₃ Molecular Weight: 230,8516 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11856025 CAS-Nummer: 889849-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...