6-Bromisatin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-1H-indole-2,3-dione, 6-Bromoindoline-2,3-dione
OR3605-25G
OR3605-5G
OR3605-100G
APOSOR3605-25GEA
62.5
EUR
APOSOR3605-25G
APOSOR3605-5G
APOSOR3605-100G
6-Bromisatin 98%
6-Bromisatin
Formel:
C₈H₄BrNO₂ Molecular Weight: 226,03 g/mol Schmelzpunkt: 274 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01631138 CAS-Nummer: 6326-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...