4-Bromisatin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromoindoline-2,3-dione

OR3606-25G OR3606-5G OR3606-100G
APOSOR3606-25GEA 37.7 EUR
APOSOR3606-25G APOSOR3606-5G APOSOR3606-100G
4-Bromisatin 98%
4-Bromisatin
Formel: C₈H₄BrNO₂
Molecular Weight: 226,0269 g/mol
Schmelzpunkt: 253…256 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03618555
CAS-Nummer: 20780-72-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR