5-Amino-2-methylphenylboronic acid pinacol ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-2-methylphenylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-(5-amino-2-methylphenyl)-1,3,2-dioxaborolane, 4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenamine

OR360650-5G
APOSOR360650-5GEA 62.5 EUR
APOSOR360650-5G
5-Amino-2-methylphenylboronic acid pinacol ester 97%
5-Amino-2-methylphenylboronic acid pinacol ester
Formel: C₁₃H₂₀BNO₂
Molecular Weight: 233,118 g/mol
Schmelzpunkt: 82…86 °C
Lagertemperatur: Kühlschrank
MDL: MFCD05663859
CAS-Nummer: 882670-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR