Benzo[b]furan-3-boronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Benzofuran-3-ylboronic acid
OR360664-5G
APOSOR360664-5GEA
148
EUR
APOSOR360664-5G
Benzo[b]furan-3-boronsäure 98%
Benzo[b]furan-3-boronsäure
Formel:
C₈H₇BO₃ Molecular Weight: 161,95 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD06801687 CAS-Nummer: 317830-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...