Thiophene-2,5-diboronic acid bis(pinacol) ester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene, Thiophene-2,5-diboronic acid Bis(pinacol) ester, 2,2'-(2,5-Thiophenediyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

OR360704-25G
APOSOR360704-25GEA 186 EUR
APOSOR360704-25G
Thiophene-2,5-diboronic acid bis(pinacol) ester 98%
Thiophene-2,5-diboronic acid bis(pinacol) ester
Formel: C₁₆H₂₆B₂O₄S
Molecular Weight: 336,06 g/mol
Schmelzpunkt: 227…231 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09800563
CAS-Nummer: 175361-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR