2,6-Dimethyl-3-pyridinboronsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
OR361168-250MG
OR361168-1G
OR361168-5G
APOSOR361168-250MGEA
92.8
EUR
APOSOR361168-250MG
APOSOR361168-1G
APOSOR361168-5G
2,6-Dimethyl-3-pyridinboronsäure 96%
2,6-Dimethyl-3-pyridineboronic acid
Formel:
C₇H₁₀BNO₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD07437925 CAS-Nummer: 693774-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...