3-Methoxy-2-naphthalinboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methoxynaphthalen-2-boronic acid, (3-Methoxynaphthalen-2-yl)boronic acid
OR361380-1G
OR361380-5G
APOSOR361380-1GEA
21.9
EUR
APOSOR361380-1G
APOSOR361380-5G
3-Methoxy-2-naphthalinboronsäure 98%
3-Methoxy-2-naphthaleneboronic acid
Formel:
C₁₁H₁₁BO₃ Molecular Weight: 202.0100 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12025968 CAS-Nummer: 104115-76-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...