Oxindol-6-boronsäure 95+%

Lieferant: Apollo Scientific

Synonyme: (2-Oxoindolin-6-yl)boronic acid

OR361513-1G
APOSOR361513-1GEA 236 EUR
APOSOR361513-1G
Oxindol-6-boronsäure 95+%
Oxindol-6-boronsäure
Formel: C₈H₈BNO₃
Molecular Weight: 176,9650 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12913936
CAS-Nummer: 1217500-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR