(E)-1-Pentenylboronic acid pinacol ester
Lieferant: Apollo Scientific
Synonyme:
(E)-1-Pentene-1-boronic Acid Pinacol Ester
OR361518-25G
OR361518-5G
OR361518-1G
APOSOR361518-25GEA
934
EUR
APOSOR361518-25G
APOSOR361518-5G
APOSOR361518-1G
(E)-1-Pentenylboronic acid pinacol ester
(E)-1-Pentenylboronic acid pinacol ester
Formel:
C₁₁H₂₁BO₂ Molecular Weight: 196,10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663882 CAS-Nummer: 161395-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...