5-Carboxy-2-methoxyphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Borono-4-methoxybenzoic acid

OR3620-1G OR3620-500MG
APOSOR3620-1GEA 267 EUR
APOSOR3620-1G APOSOR3620-500MG
5-Carboxy-2-methoxyphenylboronsäure 98%
5-Carboxy-2-methoxyphenylboronsäure
Formel: C₈H₉BO₅
Molecular Weight: 195,9700 g/mol
Schmelzpunkt: 207…210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06203261
CAS-Nummer: 730971-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR