Fmoc-Cys(StBu)-OH

Lieferant: Apollo Scientific

Synonyme: N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-S-(tert-butylthio)-L-cysteine

OR36369-25G
APOSOR36369-25GEA 632 EUR
APOSOR36369-25G
Fmoc-Cys(StBu)-OH
Fmoc-Cys(StBu)-OH
Formel: C₂₂H₂₅NO₄S₂
Molecular Weight: 431,565 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00042617
CAS-Nummer: 73724-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR