7-Chlorindol

Lieferant: Apollo Scientific
Achtung

OR3666-1G
APOSOR3666-1GEA 34.8 EUR
APOSOR3666-1G
7-Chlorindol
7-Chlorindol
Formel: C₈H₆ClN
Molecular Weight: 151,6 g/mol
Schmelzpunkt: 57…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01321301
CAS-Nummer: 53924-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR