1-Methyl-4-nitro-1H-indazol

Lieferant: Apollo Scientific
Achtung

OR3692-1G OR3692-250MG OR3692-500MG
APOSOR3692-1GEA 36.4 EUR
APOSOR3692-1G APOSOR3692-250MG APOSOR3692-500MG
1-Methyl-4-nitro-1H-indazol
1-Methyl-4-nitro-1H-indazole
Formel: C₈H₇N₃O₂
Molecular Weight: 177.1600 g/mol
Schmelzpunkt: 141…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607929
CAS-Nummer: 26120-43-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR