4-Azaindol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H-Pyrrolo[2,3-e]pyridin
OR3693-1G
OR3693-5G
OR3693-25G
APOSOR3693-1GEA
21.9
EUR
APOSOR3693-1G
APOSOR3693-5G
APOSOR3693-25G
4-Azaindol
4-Azaindol
Formel:
C₇H₆N₂ Molecular Weight: 118.14 g/mol Schmelzpunkt: 125…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00971977 CAS-Nummer: 272-49-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...