Ethyl-3,4,5-trimethoxybenzoylacetate 95+%
Lieferant: Apollo Scientific
OR3697-25G
OR3697-5G
OR3697-1G
APOSOR3697-25GEA
845
EUR
APOSOR3697-25G
APOSOR3697-5G
APOSOR3697-1G
Ethyl-3,4,5-trimethoxybenzoylacetate 95+%
Ethyl-3,4,5-trimethoxybenzoylacetate
Formel:
C₁₄H₁₈O₆ Molecular Weight: 282,292 g/mol Schmelzpunkt: 87…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00026908 CAS-Nummer: 3044-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...