5-Chlor-2-methylindol
Lieferant: Apollo Scientific
OR3699-5G
APOSOR3699-5GEA
56.7
EUR
APOSOR3699-5G
5-Chlor-2-methylindol
5-Chloro-2-methylindole
Formel:
C₉H₈ClN Molecular Weight: 165,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005619 CAS-Nummer: 1075-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...