ω-Brom-2-cyanacetophenon 95%
Lieferant: Apollo Scientific
Synonyme:
2-Brom-2'-cyanacetophenon, 2-Cyanphenacylbromid, α-Brom-2-cyanacetophenon, 2-(Bromacetyl)benzonitril, Brommethyl-o-cyanphenylketon
OR370138-1G
OR370138-250MG
APOSOR370138-1GEA
928
EUR
APOSOR370138-1G
APOSOR370138-250MG
ω-Brom-2-cyanacetophenon 95%
ω-Brom-2-cyanacetophenon
Formel:
C₉H₆BrNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407832 CAS-Nummer: 683274-86-4 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...