2-Brom-5-chlor-4-pyridinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-5-chloropyridine-4-boronic acid, (2-Bromo-5-chloropyridin-4-yl)boronic acid

OR3704-1G OR3704-250MG OR3704-5G
APOSOR3704-1GEA 91.5 EUR
APOSOR3704-1G APOSOR3704-250MG APOSOR3704-5G
2-Brom-5-chlor-4-pyridinboronsäure
2-Bromo-5-chloro-4-pyridineboronic acid
Formel: C₅H₄BBrClNO₂
Molecular Weight: 236,26 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08064041
CAS-Nummer: 1072952-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR