4-Brom-2-thenoesäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-thiophenecarboxylic acid

OR3714-100G OR3714-25G OR3714-5G
APOSOR3714-100GEA 201 EUR
APOSOR3714-100G APOSOR3714-25G APOSOR3714-5G
4-Brom-2-thenoesäure 98%
4-Brom-2-thenoesäure
Formel: C₅H₃BrO₂S
Molecular Weight: 207.05 g/mol
Schmelzpunkt: 118…122 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03422294
CAS-Nummer: 16694-18-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR