3-Amino-2-phenylpyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Phenylpyridin-3-amine
OR3718-250MG
OR3718-5G
APOSOR3718-250MGEA
63.8
EUR
APOSOR3718-250MG
APOSOR3718-5G
3-Amino-2-phenylpyridine
3-Amino-2-phenylpyridine
Formel:
C₁₁H₁₀N₂ Molecular Weight: 170.22 g/mol Schmelzpunkt: 62…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114113 CAS-Nummer: 101601-80-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...