2-Amino-4-chlorpyrimidin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-chloropyrimidin-2-amine
OR3722-100G
OR3722-25G
OR3722-5G
APOSOR3722-100GEA
316
EUR
APOSOR3722-100G
APOSOR3722-25G
APOSOR3722-5G
2-Amino-4-chlorpyrimidin 98%
2-Amino-4-chloropyrimidine
Formel:
C₄H₄ClN₃ Molecular Weight: 129,55 g/mol Schmelzpunkt: 155…160 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00038021 CAS-Nummer: 3993-78-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...