2-Bromo-3,5-dinitrobenzoic acid 98%

Lieferant: Apollo Scientific
Achtung

OR3735-100G OR3735-25G OR3735-5G
APOSOR3735-100GEA 435 EUR
APOSOR3735-100G APOSOR3735-25G APOSOR3735-5G
2-Bromo-3,5-dinitrobenzoic acid 98%
2-Bromo-3,5-dinitrobenzoic acid
Formel: C₇H₃BrN₂O₆
Schmelzpunkt: 213…216 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00458630
CAS-Nummer: 116529-60-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR