Ethyl-3-brom-5-nitrobenzoat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Brom-5-nitrobenzoesäureethylester

OR3749-1G OR3749-5G OR3749-25G
APOSOR3749-1GEA 65.4 EUR
APOSOR3749-1G APOSOR3749-5G APOSOR3749-25G
Ethyl-3-brom-5-nitrobenzoat 98%
Ethyl-3-brom-5-nitrobenzoat
Formel: C₉H₈BrNO₄
Molecular Weight: 274,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258760
CAS-Nummer: 690260-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR