Ethyl-4-oxazolcarboxylat 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl oxazole-4-carboxylate
OR3767-1G
OR3767-5G
OR3767-25G
APOSOR3767-1GEA
21.9
EUR
APOSOR3767-1G
APOSOR3767-5G
APOSOR3767-25G
Ethyl-4-oxazolcarboxylat 97%
Ethyl-4-oxazolcarboxylat
Formel:
C₆H₇NO₃ Molecular Weight: 141,1300 g/mol Schmelzpunkt: 48 °C Dichte: 1,177 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114940 CAS-Nummer: 23012-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...