5-Brom-2-hydrazinopyridin 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (5-Bromo-2-pyridyl)hydrazine, 5-Bromo-2-hydrazinopyridine

OR3769-100G OR3769-25G OR3769-5G
APOSOR3769-100GEA 154 EUR
APOSOR3769-100G APOSOR3769-25G APOSOR3769-5G
5-Brom-2-hydrazinopyridin 98%
(5-Bromopyridin-2-yl)hydrazine
Formel: C₅H₆BrN₃
Molecular Weight: 188.0252 g/mol
Schmelzpunkt: 132…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00513424
CAS-Nummer: 77992-44-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR