Methyl-2-amino-5-bromisonicotinat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-amino-5-bromo-4-pyridinecarboxylate

OR3789-1G OR3789-25G OR3789-5G
APOSOR3789-1GEA 21.9 EUR
APOSOR3789-1G APOSOR3789-25G APOSOR3789-5G
Methyl-2-amino-5-bromisonicotinat 98%
Methyl-2-amino-5-bromisonicotinat
Formel: C₇H₇BrN₂O₂
Molecular Weight: 231,0467 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09027294
CAS-Nummer: 882499-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR