4-Piperidin-1-ylmethylbenzylamine 97%

Lieferant: Apollo Scientific

OR3841-1G OR3841-5G
APOSOR3841-1GEA 340 EUR
APOSOR3841-1G APOSOR3841-5G
4-Piperidin-1-ylmethylbenzylamine 97%
4-Piperidin-1-ylmethylbenzylamine
Formel: C₁₃H₂₀N₂
Molecular Weight: 204,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07345530
CAS-Nummer: 91271-81-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR