1,3-Benzodioxol-4-carbonsäure 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzo[d][1,3]dioxole-4-carboxylic acid

OR3878-1G OR3878-250MG
APOSOR3878-1GEA 87 EUR
APOSOR3878-1G APOSOR3878-250MG
1,3-Benzodioxol-4-carbonsäure 97+%
1,3-Benzodioxol-4-carbonsäure
Formel: C₈H₆O₄
Molecular Weight: 166,1308 g/mol
Schmelzpunkt: 224…225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01076411
CAS-Nummer: 5768-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR