2-Morpholin-4-ylisonicotinic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR3881-100MG
OR3881-1G
APOSOR3881-100MGEA
46.4
EUR
APOSOR3881-100MG
APOSOR3881-1G
2-Morpholin-4-ylisonicotinic acid 97%
2-Morpholin-4-ylisonicotinic acid
Formel:
C₁₀H₁₂N₂O₃ Schmelzpunkt: 191…193 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411666 CAS-Nummer: 295349-64-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...