6-[4-(tert-Butoxycarbonyl)piperazino]nicotinic acid

Lieferant: Apollo Scientific
Achtung

OR3882-1G OR3882-250MG
APOSOR3882-1GEA 260 EUR
APOSOR3882-1G APOSOR3882-250MG
6-[4-(tert-Butoxycarbonyl)piperazino]nicotinic acid
6-[4-(tert-Butoxycarbonyl)piperazino]nicotinic acid
Formel: C₁₅H₂₁N₃O₄
Schmelzpunkt: 298…300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06245543
CAS-Nummer: 201809-22-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR