2-Desoxy-D-glucose

Lieferant: Apollo Scientific
Achtung

Synonyme: (3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal

OR3900T-5G
APOSOR3900T-5GEA 50.9 EUR
APOSOR3900T-5G
2-Desoxy-D-glucose
2-Deoxy-D-glucose
Formel: C₆H₁₂O₅
Molecular Weight: 164,16 g/mol
Schmelzpunkt: 147…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007013
CAS-Nummer: 154-17-6
EINECS: 205-823-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR