5-Bromnicotinonitril 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Brom-5-pyridincarbonitril, 5-Bromnicotinsäurenitril, 3-Brom-5-pyridincarbonsäurenitril, 3-Brom-5-cyanpyridin

OR3918-5G
APOSOR3918-5GEA 21.9 EUR
APOSOR3918-5G
5-Bromnicotinonitril 97%
5-Bromnicotinonitril
Formel: C₆H₃BrN₂
Molecular Weight: 183,01 g/mol
Schmelzpunkt: 103…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00174363
CAS-Nummer: 35590-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR