7-Nitro-1H-indole-2-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
CRT0044876
OR3926-1G
OR3926-5G
APOSOR3926-1GEA
72.5
EUR
APOSOR3926-1G
APOSOR3926-5G
7-Nitro-1H-indole-2-carboxylic acid 97%
7-Nitro-1H-indole-2-carboxylic acid
Formel:
C₉H₆N₂O₄ Schmelzpunkt: 264…266 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00044720 CAS-Nummer: 6960-45-8 EINECS: 230-154-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...